siRNA Details
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 8 | 1292 | ||
NM_031921.5 | NP_114127.3 | 10 | 1175 | |||
XM_005244806.3 | XP_005244863.1 | 10 | 1198 | |||
XM_011542241.2 | XP_011540543.1 | 10 | 1198 | |||
XM_011542243.2 | XP_011540545.1 | 9 | 911 | |||
XM_011542244.1 | XP_011540546.1 | 10 | 1342 | |||
GenBank mRNA | AB033099.1 | - | 8 | 1381 | ||
AK024028.1 | - | 8 | 760 | |||
AK127857.1 | - | 3 | 1826 | |||
AK290586.1 | - | 10 | 1167 | |||
AL834179.1 | - | 2 | 1013 | |||
BC002542.2 | - | 10 | 1101 | 3138578 | ||
BC018701.1 | - | 10 | 1101 | 3139886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
