siRNA Details
Gene Symbol: FAM177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136691.1 | 5 | 553 | |||
XM_017001283.1 | XP_016856772.1 | 4 | 327 | |||
XM_017001284.1 | XP_016856773.1 | 4 | 327 | |||
GenBank mRNA | AK125494.1 | - | 5 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
