siRNA Details
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002363.4 | NP_002354.2 | 1 | 50 | ||
NM_177415.2 | NP_803134.1 | 1 | 50 | |||
GenBank mRNA | X82539.1 | - | 1 | 51 |
Gene Symbol: MAGEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002367.3 | NP_002358.1 | 1 | 1841 | ||
GenBank mRNA | BC032852.2 | - | 1 | 1729 | 5267183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
