siRNA Details
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 1062 | ||
NM_001177997.1 | NP_001171468.1 | 3 | 955 | |||
NM_144671.4 | NP_653272.2 | 3 | 974 | |||
XM_006719257.3 | XP_006719320.1 | 4 | 1307 | |||
XM_011537976.2 | XP_011536278.1 | 2 | 781 | |||
XM_017018883.1 | XP_016874372.1 | 2 | 803 | |||
GenBank mRNA | AK056918.1 | - | 3 | 360 | ||
AK093052.1 | - | 2 | 678 | |||
AK299344.1 | - | 3 | 924 | |||
BC034809.1 | - | 3 | 875 | 4943538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
