siRNA Details
Gene Symbol: TIFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052864.2 | NP_443096.1 | 1, 2 | 272 | ||
GenBank mRNA | AB097011.1 | - | 1, 2 | 274 | ||
AB097038.1 | - | 1, 2 | 274 | |||
AK292900.1 | - | 1, 2 | 269 | |||
BC008294.1 | - | 1, 2 | 256 | |||
BC014259.2 | - | 1, 2 | 252 | 4763971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
