siRNA Details
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017623.4 | NP_060093.3 | 2 | 1286 | ||
XM_011510957.2 | XP_011509259.1 | 2 | 1646 | |||
XM_017003800.1 | XP_016859289.1 | 2 | 1646 | |||
GenBank mRNA | AF216965.1 | - | 2 | 385 | ||
AK126847.1 | - | 2 | 1286 | |||
BC018604.1 | - | 1 | 31 | |||
BC037272.1 | - | 2 | 1295 | 4823510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
