siRNA Details
Gene Symbol: TIFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052864.2 | NP_443096.1 | 2 | 421 | ||
GenBank mRNA | AB062110.1 | - | 1 | 139 | ||
AB097011.1 | - | 2 | 423 | |||
AB097038.1 | - | 2 | 423 | |||
AK292900.1 | - | 2 | 418 | |||
BC008294.1 | - | 2 | 405 | |||
BC014259.2 | - | 2 | 401 | 4763971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
