siRNA Details
Gene Symbol: ZC3H11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319238.1 | NP_001306167.1 | 2 | 434 | ||
NM_001319239.1 | NP_001306168.1 | 2 | 434 | |||
NM_014827.5 | NP_055642.3 | 2 | 415 | |||
XM_005245643.2 | XP_005245700.1 | 2 | 434 | |||
XM_006711673.2 | XP_006711736.1 | 2 | 176 | |||
XM_011510201.1 | XP_011508503.1 | 2 | 304 | |||
XM_017002956.1 | XP_016858445.1 | 2 | 307 | |||
XM_017002957.1 | XP_016858446.1 | 2 | 159 | |||
XM_017002958.1 | XP_016858447.1 | 2 | 146 | |||
XM_017002959.1 | XP_016858448.1 | 2 | 159 | |||
XM_017002960.1 | XP_016858449.1 | 2 | 307 | |||
XM_017002963.1 | XP_016858452.1 | 2 | 298 | |||
GenBank mRNA | AK095858.1 | - | 2 | 301 | ||
BC014268.2 | - | 2 | 253 | 3844154 | ||
BC038513.2 | - | 2 | 265 | |||
DA800934.1 | - | 2 | 329 | |||
DB066230.1 | - | 2 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
