siRNA Details
Gene Symbol: LOC441155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271675.1 | NP_001258604.1 | 2 | 812 | ||
XM_017010889.1 | XP_016866378.1 | 2 | 328 | |||
GenBank mRNA | AK302514.1 | - | 2 | 768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
