siRNA Details
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 10, 11 | 1202 | ||
XM_005255318.1 | XP_005255375.1 | 10, 11 | 1196 | |||
GenBank mRNA | AU132793.1 | - | 3, 4 | 264 | ||
BM507095.1 | - | 1, 2 | 70 | |||
BQ954152.1 | - | 404 | ||||
CD653707.1 | - | 4, 5 | 488 | |||
DA392676.1 | - | 5, 6 | 531 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 10, 11 | 1133 | ||
GenBank mRNA | BC065535.1 | - | 10, 11 | 1086 | 6007603 | |
X57398.1 | - | 10, 11 | 971 |
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 10, 11 | 1139 | ||
NM_173614.2 | NP_775885.1 | 10, 11 | 1139 | |||
XM_017023156.1 | XP_016878645.1 | 10, 11 | 1186 | |||
GenBank mRNA | AK225803.1 | - | 10, 11 | 1109 | ||
AL832855.1 | - | 10, 11 | 1133 | |||
BC028389.1 | - | 9, 10 | 999 | 4824724 | ||
BC041131.1 | - | 10, 11 | 1113 | 5757234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
