siRNA Details
Gene Symbol: SRSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006275.5 | NP_006266.2 | 1 | 197 | ||
NR_034009.1 | 1 | 197 | ||||
GenBank mRNA | AB208976.1 | - | 1 | 156 | ||
AB451251.1 | - | 1 | 27 | |||
AB451375.1 | - | 1 | 27 | |||
AK291203.1 | - | 1 | 165 | |||
AK300411.1 | - | 1 | 165 | |||
BC006832.2 | - | 1 | 151 | 3449841 | ||
BM788391.1 | - | 1 | 183 | |||
DC306805.1 | - | 1 | 197 | |||
EU668347.1 | - | 1 | 132 | |||
U30828.1 | - | 1 | 132 | |||
U30883.1 | - | 1 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
