siRNA Details
Gene Symbol: LOC107985911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005465.1 | XP_016860954.1 | 2 | 432 |
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027354.2 | 2 | 193 | |||
NR_027355.2 | 2 | 93 | ||||
NR_027356.2 | 2 | 93 | ||||
GenBank mRNA | AK311729.1 | - | 2 | 193 | ||
AW954772.1 | - | 2 | 82 | |||
BE814857.1 | - | 2 | 85 | |||
BG716766.1 | - | 2 | 85 | |||
BQ438821.1 | - | 2 | 94 | |||
DA808118.1 | - | 2 | 95 | |||
DB496177.1 | - | 2 | 80 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027238.1 | 2 | 61 | |||
GenBank mRNA | BC030764.1 | - | 2 | 85 | ||
BF031534.1 | - | 2 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
