siRNA Details
Gene Symbol: FAM21EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038275.2 | 5, 6 | 1758 | |||
GenBank mRNA | AK056375.1 | - | 5, 6 | 1758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
