siRNA Details
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 2, 3 | 668 | |||
NR_047527.1 | 3, 4 | 778 | ||||
GenBank mRNA | AF523265.1 | - | 2, 3 | 763 | ||
AK307215.1 | - | 2, 3 | 399 | |||
AY098593.1 | - | 2, 3 | 659 | |||
BC112977.1 | - | 2, 3 | 666 | |||
BC112978.1 | - | 2, 3 | 666 | |||
DB501751.1 | - | 3, 4 | 319 |
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002009.3 | NP_002000.1 | 3, 4 | 925 | ||
GenBank mRNA | AK313160.1 | - | 3, 4 | 843 | ||
CR542002.1 | - | 2, 3 | 396 | |||
M60828.1 | - | 3, 4 | 841 | |||
S81661.1 | - | 3, 4 | 841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
