siRNA Details
Gene Symbol: OR2T34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001821.1 | NP_001001821.1 | 1 | 115 | ||
GenBank mRNA | BC136924.1 | - | 1 | 137 | 9020916 | |
BC136926.1 | - | 1 | 137 | 9020918 |
Gene Symbol: OR2T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005495.1 | NP_001005495.1 | 1 | 115 | ||
GenBank mRNA | BC136915.1 | - | 1 | 140 | 9020906 | |
BC136916.1 | - | 1 | 140 | 9020907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
