siRNA Details
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 2 | 263 | |||
GenBank mRNA | AW500063.1 | - | 2 | 196 | ||
BC104473.1 | - | 2 | 263 | |||
CF141910.1 | - | 2 | 269 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 4 | 412 | |||
GenBank mRNA | AK290693.1 | - | 4 | 379 | ||
BC147000.1 | - | 4 | 366 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 4 | 553 | |||
GenBank mRNA | BC034017.1 | - | 4 | 554 | ||
BC067351.1 | - | 2 | 251 | |||
BC140728.1 | - | 3 | 376 | |||
BC171739.1 | - | 3 | 376 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027386.2 | 4 | 531 | |||
GenBank mRNA | AK289965.1 | - | 3 | 503 | ||
BT006760.1 | - | 2 | 206 | |||
DT218991.1 | - | 4 | 451 | |||
U21555.1 | - | LIT2100 5G4 pseudogene |
4 | 442 | ||
X83299.1 | - | 4 | 482 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 2 | 200 | |||
NR_133000.1 | 2 | 200 | ||||
GenBank mRNA | AW955360.1 | - | 1 | 28 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 4 | 440 | |||
NR_132997.1 | 6 | 699 | ||||
NR_132998.1 | 3 | 367 | ||||
GenBank mRNA | BC144584.1 | - | 4 | 376 | ||
BC144585.1 | - | 3 | 303 | |||
BM553931.1 | - | 6 | 699 |
Gene Symbol: LOC100133050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027503.1 | 2 | 183 | |||
GenBank mRNA | BI830432.1 | - | 2 | 182 | ||
BQ109458.1 | - | 2 | 183 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 1 | 157 | |||
NR_029426.1 | 4 | 449 | ||||
GenBank mRNA | AK094021.1 | - | 5 | 854 | ||
AK097091.1 | - | 5 | 829 | |||
AK289832.1 | - | 4 | 531 | |||
AK315849.1 | - | 4 | 522 | |||
BC131740.1 | - | 2 | 263 | |||
X75940.1 | - | 1 | 158 | |||
X83300.1 | - | 2 | 272 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 5 | 525 | |||
GenBank mRNA | AK289851.1 | - | 4 | 451 | ||
CN312642.1 | - | 2 | 315 | |||
X83301.1 | - | 5 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
