siRNA Details
Gene Symbol: LOC101927284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110754.1 | 4 | 368 |
Gene Symbol: EPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749375.1 | - | 8 | 6885 |
Gene Symbol: LOC100131372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126634.1 | - | 1 | 3463 |
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308843.1 | - | 4 | 487 |
Gene Symbol: ZNF527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032453.1 | NP_115829.1 | 5 | 4535 | ||
XM_005259328.4 | XP_005259385.1 | 5 | 4627 | |||
XM_005259329.3 | XP_005259386.1 | 4 | 4541 | |||
XM_017027380.1 | XP_016882869.1 | 3 | 4516 | |||
XM_017027381.1 | XP_016882870.1 | 4 | 4527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
