siRNA Details
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000694.3 | NP_000685.1 | 6 | 634 | ||
NM_001030010.2 | NP_001025181.1 | 5 | 523 | |||
NM_001161473.2 | NP_001154945.1 | 6 | 692 | |||
GenBank mRNA | AB209651.1 | - | 4 | 2789 | ||
AK291505.1 | - | 6 | 617 | |||
BC002553.2 | - | 6 | 616 | |||
BC013584.2 | - | 6 | 587 | 3878124 | ||
BC033099.1 | - | 5 | 461 | 5446816 | ||
BM561843.1 | - | 6 | 633 | |||
BT009832.1 | - | 5 | 545 | |||
EF411198.1 | - | 6 | 661 | |||
U10868.1 | - | 6 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
