siRNA Details
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000694.3 | NP_000685.1 | 5, 6 | 565 | ||
NM_001030010.2 | NP_001025181.1 | 4, 5 | 454 | |||
NM_001161473.2 | NP_001154945.1 | 5, 6 | 623 | |||
GenBank mRNA | AB209651.1 | - | 3, 4 | 2720 | ||
AK291505.1 | - | 5, 6 | 548 | |||
BC002553.2 | - | 5, 6 | 547 | |||
BC013584.2 | - | 5, 6 | 518 | 3878124 | ||
BC033099.1 | - | 4, 5 | 392 | 5446816 | ||
BM561843.1 | - | 5, 6 | 564 | |||
BT009832.1 | - | 4, 5 | 476 | |||
EF411198.1 | - | 5, 6 | 592 | |||
U10868.1 | - | 5, 6 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
