siRNA Details
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 5 | 467 | ||
XM_005260421.2 | XP_005260478.1 | 2 | 77 | |||
XM_011528843.1 | XP_011527145.1 | 5 | 456 | |||
XM_011528844.1 | XP_011527146.1 | 2 | 139 | |||
XM_017027864.1 | XP_016883353.1 | 6 | 549 | |||
XM_017027865.1 | XP_016883354.1 | 6 | 750 | |||
XM_017027866.1 | XP_016883355.1 | 5 | 670 | |||
XM_017027867.1 | XP_016883356.1 | 5 | 505 | |||
XM_017027871.1 | XP_016883360.1 | 6 | 550 | |||
GenBank mRNA | AF220416.1 | - | 5 | 431 | ||
AF258787.1 | - | 5 | 453 | |||
AF348207.1 | - | 4 | 392 | |||
AK054597.1 | - | 6 | 724 | |||
AK314503.1 | - | 5 | 431 | |||
AL558423.3 | - | 4 | 390 | |||
AY027523.1 | - | 4 | 392 | |||
AY134747.1 | - | 5 | 444 | |||
BC016948.1 | - | 5 | 420 | |||
BC048210.1 | - | 6 | 500 | |||
BC080598.1 | - | 5 | 431 | |||
BC093405.1 | - | 5 | 467 | |||
BC150178.1 | - | 5 | 433 | 40148074 | ||
BG778940.1 | - | 6 | 537 | |||
BM906582.1 | - | 5 | 456 | |||
BX643875.1 | - | 5 | 425 | |||
CD690482.1 | - | 5 | 469 | |||
L09749.1 | - | 4 | 349 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172674, AK038573, BC060121, AK033017, AK045309.
Gene Ontology Categories:
