siRNA Details
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 5 | 492 | ||
XM_005260421.2 | XP_005260478.1 | 2 | 102 | |||
XM_011528843.1 | XP_011527145.1 | 5 | 481 | |||
XM_011528844.1 | XP_011527146.1 | 2 | 164 | |||
XM_017027864.1 | XP_016883353.1 | 6 | 574 | |||
XM_017027865.1 | XP_016883354.1 | 6 | 775 | |||
XM_017027866.1 | XP_016883355.1 | 5 | 695 | |||
XM_017027867.1 | XP_016883356.1 | 5 | 530 | |||
XM_017027871.1 | XP_016883360.1 | 6 | 575 | |||
GenBank mRNA | AF220416.1 | - | 5 | 456 | ||
AF258787.1 | - | 5 | 478 | |||
AF348207.1 | - | 4 | 417 | |||
AK054597.1 | - | 6 | 749 | |||
AK314503.1 | - | 5 | 456 | |||
AL558423.3 | - | 4 | 415 | |||
AY027523.1 | - | 4 | 417 | |||
AY134747.1 | - | 5 | 469 | |||
BC016948.1 | - | 5 | 445 | |||
BC048210.1 | - | 6 | 525 | |||
BC080598.1 | - | 5 | 456 | |||
BC093405.1 | - | 5 | 492 | |||
BC150178.1 | - | 5 | 458 | 40148074 | ||
BG778940.1 | - | 6 | 562 | |||
BM906582.1 | - | 5 | 481 | |||
BX643875.1 | - | 5 | 450 | |||
CD690482.1 | - | 5 | 494 | |||
L09749.1 | - | 4 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
