siRNA Details
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 2 | 2381 | ||
XM_017012623.1 | XP_016868112.1 | 2 | 2436 | |||
GenBank mRNA | AK090570.1 | - | 1 | 24 | ||
AK123596.1 | - | 1 | 580 | |||
AK160365.1 | - | 2 | 2437 | |||
BC136661.1 | - | 2 | 2295 | 9020651 | ||
BX649178.1 | - | 1 | 266 | |||
CR749203.1 | - | 2 | 2321 |
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 1 | 1300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
