siRNA Details
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016026.3 | NP_057110.3 | 5 | 606 | ||
GenBank mRNA | AB209223.1 | - | 1 | 2493 | ||
AF151840.1 | - | 5 | 556 | |||
AF167438.1 | - | 5 | 556 | |||
AF395068.1 | - | 5 | 556 | |||
AK057195.1 | - | 5 | 442 | |||
AK074749.1 | - | 5 | 586 | |||
AK289427.1 | - | 5 | 504 | |||
AK307500.1 | - | 6 | 503 | |||
AK314465.1 | - | 5 | 572 | |||
BC000112.1 | - | 5 | 523 | 3504920 | ||
BC011727.2 | - | 5 | 521 | 4135182 | ||
BC026274.1 | - | 5 | 543 | 4250934 | ||
BC037302.1 | - | 5 | 583 | 5261204 | ||
BC051291.1 | - | 5 | 516 | 3533223 | ||
CR457180.1 | - | 5 | 516 | |||
KU178627.1 | - | 5 | 516 | |||
KU178628.1 | - | 5 | 516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
