siRNA Details
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252650.1 | NP_001239579.1 | 6 | 940 | ||
NM_016026.3 | NP_057110.3 | 7 | 1150 | |||
GenBank mRNA | AB209223.1 | - | 4 | 3211 | ||
AF151840.1 | - | 7 | 1099 | |||
AF167438.1 | - | 7 | 1100 | |||
AK057195.1 | - | 7 | 986 | |||
AK074749.1 | - | 7 | 1130 | |||
AK289427.1 | - | 7 | 1048 | |||
AK293355.1 | - | 6 | 880 | |||
BC000112.1 | - | 7 | 1067 | 3504920 | ||
BC011727.2 | - | 7 | 1065 | 4135182 | ||
BC026274.1 | - | 7 | 1087 | 4250934 | ||
BC037302.1 | - | 7 | 1127 | 5261204 | ||
BC051291.1 | - | 7 | 1060 | 3533223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
