siRNA Details
Gene Symbol: OPN1LW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020061.5 | NP_064445.2 | 2 | 250 | ||
GenBank mRNA | BQ639996.1 | - | 2 | 193 | ||
CR749814.1 | - | 2 | 208 | |||
DN693335.1 | - | 2 | 250 | |||
EL952084.1 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
