siRNA Details
Gene Symbol: CHRAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017444.5 | NP_059140.1 | 3 | 488 | ||
NR_023360.2 | 2 | 361 | ||||
NR_040712.1 | 3 | 183 | ||||
GenBank mRNA | AF226076.1 | - | 3 | 286 | ||
AK023537.1 | - | 3 | 481 | |||
AK308709.1 | - | 2 | 231 | |||
AK311595.1 | - | 2 | 879 | |||
AL543044.3 | - | 2 | 348 | |||
BC015891.1 | - | 3 | 325 | 3908717 | ||
BU160855.1 | - | 3 | 186 | |||
DA647606.1 | - | 3 | 488 | |||
DA887467.1 | - | 3 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
