siRNA Details
Gene Symbol: CHRAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017444.5 | NP_059140.1 | 3 | 512 | ||
NR_023360.2 | 2 | 385 | ||||
NR_040712.1 | 3 | 207 | ||||
GenBank mRNA | AF226076.1 | - | 3 | 310 | ||
AK023537.1 | - | 3 | 505 | |||
AK308709.1 | - | 2 | 255 | |||
AK311595.1 | - | 2 | 903 | |||
AL543044.3 | - | 2 | 372 | |||
BC015891.1 | - | 3 | 349 | 3908717 | ||
BU160855.1 | - | 3 | 210 | |||
DA647606.1 | - | 3 | 512 | |||
DA887467.1 | - | 3 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
