siRNA Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019726.1 | XP_016875215.1 | 1 | 395 | ||
XM_017019727.1 | XP_016875216.1 | 1 | 395 | |||
XM_017019728.1 | XP_016875217.1 | 1 | 395 | |||
XM_017019729.1 | XP_016875218.1 | 1 | 395 | |||
XM_017019730.1 | XP_016875219.1 | 1 | 395 | |||
XM_017019731.1 | XP_016875220.1 | 1 | 395 | |||
XM_017019732.1 | XP_016875221.1 | 1 | 395 | |||
XM_017019733.1 | XP_016875222.1 | 1 | 395 | |||
XM_017019735.1 | XP_016875224.1 | 1 | 395 | |||
XM_017019736.1 | XP_016875225.1 | 1 | 395 | |||
XM_017019737.1 | XP_016875226.1 | 1 | 395 | |||
XM_017019738.1 | XP_016875227.1 | 1 | 395 | |||
XM_017019739.1 | XP_016875228.1 | 1 | 395 | |||
XM_017019740.1 | XP_016875229.1 | 1 | 395 | |||
XM_017019741.1 | XP_016875230.1 | 1 | 395 | |||
XM_017019742.1 | XP_016875231.1 | 1 | 395 | |||
GenBank mRNA | AK122964.1 | - | 1 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
