siRNA Details
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020661.2 | NP_065712.1 | 2 | 116 | ||
XM_011520772.2 | XP_011519074.1 | 2 | 235 | |||
XM_011520773.2 | XP_011519075.1 | 2 | 240 | |||
GenBank mRNA | AB040431.1 | - | 2 | 113 | ||
AF529815.1 | - | 1 | 37 | |||
AF529816.1 | - | 1 | 37 | |||
AF529817.1 | - | 1 | 37 | |||
AF529818.1 | - | 1 | 37 | |||
AF529819.1 | - | 1 | 37 | |||
AF529820.1 | - | 1 | 37 | |||
AF529821.1 | - | 1 | 37 | |||
AF529822.1 | - | 1 | 37 | |||
AF529823.1 | - | 1 | 37 | |||
AF529824.1 | - | 1 | 37 | |||
AF529825.1 | - | 1 | 37 | |||
AF529826.1 | - | 1 | 37 | |||
AF529827.1 | - | 1 | 37 | |||
AY534975.1 | - | 1 | 37 | |||
AY536516.1 | - | 1 | 37 | |||
AY536517.1 | - | 1 | 37 | |||
AY541058.1 | - | 1 | 37 | |||
BC006296.2 | - | 2 | 113 | 4054915 | ||
BT007402.1 | - | 1 | 37 | |||
BX402063.2 | - | 2 | 130 | |||
DQ431660.1 | - | 1 | 31 | |||
EU794587.1 | - | 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
