siRNA Details
Gene Symbol: KISS1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032551.4 | NP_115940.2 | 5 | 1612 | ||
GenBank mRNA | AY253982.1 | - | 5 | 1596 | ||
BC141812.1 | - | 5 | 1612 | 40147721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
