siRNA Details
Gene Symbol: GPR68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177676.1 | NP_001171147.1 | 2 | 258 | ||
NM_003485.3 | NP_003476.3 | 2 | 212 | |||
XM_005268110.4 | XP_005268167.1 | 2 | 1398 | |||
XM_005268111.3 | XP_005268168.1 | 2 | 178 | |||
XM_005268112.3 | XP_005268169.1 | 2 | 118 | |||
XM_006720262.3 | XP_006720325.1 | 2 | 144 | |||
XM_011537196.2 | XP_011535498.1 | 3 | 262 | |||
XM_011537197.2 | XP_011535499.1 | 2 | 542 | |||
XM_011537198.2 | XP_011535500.1 | 2 | 768 | |||
XM_011537199.2 | XP_011535501.1 | 2 | 213 | |||
GenBank mRNA | BC098567.1 | - | 2 | 155 | 30925589 | |
BC128536.1 | - | 2 | 266 | 40120393 | ||
BM723705.1 | - | 2 | 257 | |||
CN287036.1 | - | 3 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
