siRNA Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322963.1 | NP_001309892.1 | 4 | 353 | ||
NM_001322964.1 | NP_001309893.1 | 3 | 172 | |||
NM_004116.4 | NP_004107.1 | 3 | 285 | |||
NM_054033.3 | NP_473374.1 | 3 | 285 | |||
NR_136536.1 | 5 | 442 | ||||
NR_136538.1 | 4 | 374 | ||||
NR_136539.1 | 3 | 172 | ||||
XM_011532699.2 | XP_011531001.1 | 3 | 234 | |||
XM_017003593.1 | XP_016859082.1 | 5 | 388 | |||
XM_017003594.1 | XP_016859083.1 | 3 | 152 | |||
XM_017003595.1 | XP_016859084.1 | 2 | 133 | |||
XM_017003596.1 | XP_016859085.1 | 4 | 379 | |||
GenBank mRNA | AF322070.1 | - | 3 | 177 | ||
BC002614.1 | - | 3 | 228 | 3161896 | ||
BF216711.1 | - | 3 | 140 | |||
BF969580.1 | - | 3 | 272 | |||
BI546013.1 | - | 3 | 250 | |||
BM802489.1 | - | 3 | 285 | |||
BU147063.1 | - | 3 | 123 | |||
BU508033.1 | - | 3 | 249 | |||
D38037.1 | - | 3 | 208 | |||
L37086.1 | - | 3 | 202 | |||
S69800.1 | - | 3 | 208 | |||
S69815.1 | - | 3 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
