siRNA Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322963.1 | NP_001309892.1 | 5 | 528 | ||
NM_001322964.1 | NP_001309893.1 | 4 | 347 | |||
NM_004116.4 | NP_004107.1 | 4 | 460 | |||
NM_054033.3 | NP_473374.1 | 4 | 505 | |||
NR_136536.1 | 6 | 662 | ||||
NR_136538.1 | 5 | 594 | ||||
NR_136539.1 | 4 | 392 | ||||
XM_017003593.1 | XP_016859082.1 | 6 | 563 | |||
XM_017003594.1 | XP_016859083.1 | 4 | 327 | |||
XM_017003595.1 | XP_016859084.1 | 3 | 308 | |||
XM_017003596.1 | XP_016859085.1 | 5 | 554 | |||
GenBank mRNA | AF322070.1 | - | 4 | 397 | ||
BC002614.1 | - | 4 | 448 | 3161896 | ||
BF216711.1 | - | 4 | 314 | |||
BF969580.1 | - | 4 | 447 | |||
BI546013.1 | - | 4 | 424 | |||
BM802489.1 | - | 4 | 505 | |||
BU147063.1 | - | 4 | 343 | |||
BU508033.1 | - | 4 | 424 | |||
D38037.1 | - | 4 | 383 | |||
L37086.1 | - | 4 | 377 | |||
S69815.1 | - | 4 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
