siRNA Details
Gene Symbol: PRLHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004248.2 | NP_004239.1 | 2 | 423 | ||
GenBank mRNA | AB015745.1 | - | 1 | 284 | ||
AK290461.1 | - | 2 | 423 | |||
BC095539.1 | - | 1 | 409 | 30706566 | ||
BC101490.1 | - | 1 | 358 | 8068996 | ||
BC101492.1 | - | 1 | 358 | 8068998 | ||
DA262254.1 | - | 2 | 449 | |||
EF577402.1 | - | 1 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
