siRNA Details
Gene Symbol: XPNPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003399.5 | NP_003390.4 | 20 | 2026 | ||
GenBank mRNA | BC126174.1 | - | 20 | 1943 | 8991890 | |
BC143901.1 | - | 20 | 1943 | 9052419 | ||
BQ224207.1 | - | 7 | 467 | |||
U90724.2 | - | 20 | 2046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
