siRNA Details
Gene Symbol: S1PR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003775.3 | NP_003766.1 | 1 | 885 | ||
GenBank mRNA | AJ000479.1 | - | 1 | 852 | ||
BC014970.1 | - | 1 | 855 | 4849349 | ||
FJ200495.1 | - | 2 | 692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
