siRNA Details
Gene Symbol: FFAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005303.2 | NP_005294.1 | 1 | 845 | ||
GenBank mRNA | BC095536.1 | - | 1 | 832 | ||
BC120943.1 | - | 1 | 832 | |||
BC120944.1 | - | 1 | 835 | 40115111 | ||
EF611308.1 | - | 1 | 835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
