siRNA Details
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006221.3 | NP_006212.1 | 3 | 422 | ||
NR_038422.2 | 4 | 475 | ||||
NR_038830.1 | 4 | 475 | ||||
XM_011528068.2 | XP_011526370.1 | 5 | 413 | |||
GenBank mRNA | AF086512.1 | - | 3 | 289 | ||
AK092970.1 | - | 2 | 2434 | |||
AK097558.1 | - | 3 | 306 | |||
AK291074.1 | - | 3 | 306 | |||
BC002899.1 | - | 3 | 311 | 3941595 | ||
BC031971.1 | - | 4 | 1319 | |||
BG912418.1 | - | 4 | 342 | |||
BI827520.1 | - | 3 | 428 | |||
BI827672.1 | - | 3 | 424 | |||
BT019331.1 | - | 3 | 284 | |||
CD675162.1 | - | 4 | 335 | |||
CR407654.1 | - | 3 | 284 | |||
U49070.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
