siRNA Details
Gene Symbol: TUBB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006088.5 | NP_006079.1 | 4 | 791 | ||
GenBank mRNA | AK000560.1 | - | 4 | 763 | ||
AK021414.1 | - | 4 | 619 | |||
AK026167.1 | - | 4 | 762 | |||
AK026594.1 | - | 4 | 763 | |||
AK026609.1 | - | 4 | 763 | |||
AK098686.1 | - | 4 | 692 | |||
AY167990.1 | - | 4 | 689 | |||
BC001911.1 | - | 4 | 726 | 3544236 | ||
BC002783.1 | - | 4 | 736 | 3614023 | ||
BC002885.2 | - | 4 | 733 | 3944735 | ||
BC004188.2 | - | 4 | 738 | 2964559 | ||
BC007889.2 | - | 4 | 727 | 4127195 | ||
BC008006.1 | - | 1 | 37 | |||
BC012835.2 | - | 4 | 738 | 3603755 | ||
BC019359.1 | - | 4 | 729 | 4024979 | ||
BC019829.1 | - | 4 | 752 | 5110038 | ||
BC024038.1 | - | 4 | 727 | 3616531 | ||
BC039175.1 | - | 4 | 736 | 4385885 | ||
BC071889.1 | - | 4 | 727 | 4905049 | ||
BX648521.1 | - | 3 | 1237 |
Gene Symbol: TUBB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177987.2 | NP_817124.1 | 4 | 689 | ||
XM_011519459.2 | XP_011517761.1 | 4 | 729 | |||
XM_011519460.2 | XP_011517762.1 | 3 | 375 | |||
XM_017016192.1 | XP_016871681.1 | 2 | 626 | |||
XM_017016193.1 | XP_016871682.1 | 2 | 446 | |||
GenBank mRNA | BC101270.2 | - | 4 | 682 | ||
BC101271.1 | - | 4 | 682 |
Gene Symbol: TUBB8P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014971.1 | - | 3 | 1306 | ||
BC139719.1 | - | 4 | 879 |
Gene Symbol: TUBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027156.1 | 5 | 1845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
