siRNA Details
Gene Symbol: DUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025248.1 | NP_001020419.1 | 4 | 635 | ||
NM_001025249.1 | NP_001020420.1 | 4 | 319 | |||
NM_001948.3 | NP_001939.1 | 3 | 363 | |||
XM_011521315.2 | XP_011519617.1 | 4 | 355 | |||
XM_017021988.1 | XP_016877477.1 | 4 | 621 | |||
GenBank mRNA | AB049113.2 | - | 3 | 298 | ||
AK000629.1 | - | 4 | 315 | |||
AK291515.1 | - | 3 | 352 | |||
AK312122.1 | - | 3 | 331 | |||
BC033645.1 | - | 3 | 391 | 5578877 | ||
BC070339.1 | - | 4 | 619 | 3998021 | ||
BC110377.1 | - | 4 | 614 | 5728970 | ||
BG505144.1 | - | 4 | 335 | |||
BG532018.1 | - | 365 | ||||
CR541720.1 | - | 3 | 279 | |||
CR541781.1 | - | 3 | 279 | |||
L11877.1 | - | 3 | 299 | |||
M89913.1 | - | 3 | 297 | |||
U31930.1 | - | 3 | 308 | |||
U62891.1 | - | 3 | 310 | |||
U90223.1 | - | 4 | 605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
