siRNA Details
Gene Symbol: DUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025248.1 | NP_001020419.1 | 5, 6 | 724 | ||
NM_001025249.1 | NP_001020420.1 | 5, 6 | 408 | |||
NM_001948.3 | NP_001939.1 | 4, 5 | 452 | |||
XM_011521315.2 | XP_011519617.1 | 5, 6 | 444 | |||
GenBank mRNA | AB049113.2 | - | 4, 5 | 387 | ||
AK000629.1 | - | 5, 6 | 404 | |||
AK291515.1 | - | 4, 5 | 441 | |||
AK298464.1 | - | 5, 6 | 700 | |||
AK312122.1 | - | 4, 5 | 420 | |||
BC033645.1 | - | 4, 5 | 480 | 5578877 | ||
BC070339.1 | - | 5, 6 | 708 | 3998021 | ||
BC110377.1 | - | 5, 6 | 703 | 5728970 | ||
CR541720.1 | - | 4, 5 | 368 | |||
CR541781.1 | - | 4, 5 | 368 | |||
L11877.1 | - | 4, 5 | 388 | |||
M89913.1 | - | 4, 5 | 386 | |||
U31930.1 | - | 4, 5 | 397 | |||
U62891.1 | - | 4, 5 | 399 | |||
U90223.1 | - | 5, 6 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
