siRNA Details
Gene Symbol: SSTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001052.2 | NP_001043.2 | 1 | 297 | ||
GenBank mRNA | BC069063.1 | - | 1 | 233 | 7216876 | |
BC117270.1 | - | 1 | 297 | 40125821 | ||
BC117272.1 | - | 1 | 297 | 40125823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
