siRNA Details
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 4 | 832 | |||
GenBank mRNA | BC046153.2 | - | 4 | 819 | ||
BC101598.1 | - | 3 | 288 | 8069104 | ||
BE938866.1 | - | 2 | 191 |
Gene Symbol: LOC100289561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242713.1 | NP_001229642.1 | 4 | 868 | ||
GenBank mRNA | BU073748.1 | - | 4 | 438 |
Gene Symbol: LOC100630923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038967.1 | 4 | 887 | |||
GenBank mRNA | AF178686.1 | - | 3 | 249 | ||
AK098276.1 | - | 5 | 697 | |||
AK298774.1 | - | 1 | 80 | |||
BX459931.2 | - | 4 | 850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
