siRNA Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204456.1 | NP_001191385.1 | 4 | 380 | ||
NM_021163.3 | NP_066986.1 | 3 | 575 | |||
GenBank mRNA | AK293069.1 | - | 4 | 380 | ||
BC136675.1 | - | 3 | 228 | 9020665 | ||
BC136676.1 | - | 3 | 228 | 9020666 |
Gene Symbol: LOC100289561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242713.1 | NP_001229642.1 | 3 | 695 | ||
GenBank mRNA | BU073748.1 | - | 3 | 265 |
Gene Symbol: LOC100630923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038967.1 | 3 | 714 | |||
GenBank mRNA | AF178686.1 | - | 2 | 76 | ||
AK098276.1 | - | 4 | 524 | |||
BX459931.2 | - | 3 | 677 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 3 | 659 | |||
GenBank mRNA | BC046153.2 | - | 3 | 646 | ||
BC101598.1 | - | 2 | 115 | 8069104 |
Gene Symbol: POM121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257190.2 | NP_001244119.1 | 2 | 592 | ||
NM_172020.4 | NP_742017.1 | 1 | 41 | |||
XM_006716196.3 | XP_006716259.1 | 2 | 592 | |||
XM_017012853.1 | XP_016868342.1 | 3 | 644 | |||
XM_017012854.1 | XP_016868343.1 | 3 | 284 | |||
GenBank mRNA | AB014518.1 | - | 2 | 562 | ||
BI544820.1 | - | 3 | 566 |
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099415.2 | NP_001092885.2 | 2 | 466 | ||
GenBank mRNA | BC082993.1 | - | 2 | 449 | ||
BC130587.1 | - | 2 | 173 | |||
BC130589.1 | - | 2 | 173 |
Gene Symbol: RBAK-RBAKDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204513.1 | NP_001191442.1 | 3 | 238 |
Gene Symbol: ZNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007088.1 | NP_001007089.1 | 4 | 365 | ||
NM_001178099.1 | NP_001171570.1 | 5 | 481 | |||
NM_006962.1 | NP_008893.1 | 5 | 459 | |||
GenBank mRNA | AK122874.1 | - | 5 | 464 | ||
AK299325.1 | - | 4 | 364 | |||
BC106875.2 | - | 4 | 329 | 40011764 | ||
BC114216.1 | - | 4 | 342 | 6069900 | ||
CD642892.1 | - | 5 | 494 | |||
CN311292.1 | - | 4 | 318 | |||
DC377274.1 | - | 5 | 481 |
Gene Symbol: ZNF300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172831.1 | NP_001166302.1 | 5 | 532 | ||
NM_001172832.1 | NP_001166303.1 | 3 | 435 | |||
NM_052860.2 | NP_443092.1 | 4 | 477 | |||
XM_011537702.2 | XP_011536004.1 | 5 | 632 | |||
GenBank mRNA | AF395541.1 | - | 4 | 323 | ||
AK095642.1 | - | 1 | 136 | |||
AK096465.1 | - | 4 | 474 | |||
AK300528.1 | - | 5 | 453 | |||
BC117248.1 | - | 4 | 226 | 40125799 | ||
DA497503.1 | - | 4 | 478 | |||
DB094933.1 | - | 3 | 428 | |||
DQ908918.1 | - | 5 | 338 |
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 3 | 379 | |||
GenBank mRNA | AK091436.1 | - | 3 | 349 | ||
AK096536.1 | - | 3 | 1534 | |||
AK097462.1 | - | 2 | 237 | |||
BC022559.1 | - | 3 | 379 | |||
BC117490.1 | - | 3 | 341 | |||
BC126474.1 | - | 3 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
