siRNA Details
Gene Symbol: MAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005265133.4 | XP_005265190.1 | 10 | 4271 | ||
XM_005265134.4 | XP_005265191.1 | 10 | 4684 | |||
XM_006713146.3 | XP_006713209.1 | 11 | 4303 | |||
XM_006713147.3 | XP_006713210.1 | 11 | 4720 | |||
XM_006713152.3 | XP_006713215.1 | 10 | 4669 | |||
XM_011533703.2 | XP_011532005.1 | 10 | 4220 | |||
XM_011533704.2 | XP_011532006.1 | 10 | 4220 | |||
XM_011533705.1 | XP_011532007.1 | 10 | 4220 | |||
XM_011533708.2 | XP_011532010.1 | 10 | 4220 | |||
XM_011533709.2 | XP_011532011.1 | 10 | 4220 | |||
XM_011533710.2 | XP_011532012.1 | 9 | 4097 | |||
XM_011533718.1 | XP_011532020.1 | 10 | 4220 | |||
XM_017006391.1 | XP_016861880.1 | 10 | 4220 | |||
XM_017006392.1 | XP_016861881.1 | 10 | 4220 | |||
XM_017006393.1 | XP_016861882.1 | 10 | 4252 | |||
XM_017006394.1 | XP_016861883.1 | 9 | 4546 | |||
GenBank mRNA | AK125245.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
