siRNA Details
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006932.2 | NP_001006933.2 | 3, 4 | 618 | ||
NM_001318936.1 | NP_001305865.1 | 4, 5 | 669 | |||
NM_001318938.1 | NP_001305867.1 | 2, 3 | 205 | |||
NM_021135.5 | NP_066958.2 | 2, 3 | 471 | |||
XM_006715549.3 | XP_006715612.1 | 3, 4 | 500 | |||
GenBank mRNA | AB209116.1 | - | 4, 5 | 432 | ||
AK095751.1 | - | 3, 4 | 578 | |||
AK295674.1 | - | 2, 3 | 279 | |||
AK310428.1 | - | 2, 3 | 198 | |||
BC002363.2 | - | 4, 5 | 431 | 2961482 | ||
DC402728.1 | - | 2, 3 | 211 | |||
KJ535087.1 | - | 3, 4 | 236 | |||
L07598.1 | - | 2, 3 | 216 | |||
X85106.1 | - | 4, 5 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
