siRNA Details
Gene Symbol: RDH16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320108.1 | NP_001307037.1 | 3 | 2404 | ||
NM_003708.4 | NP_003699.3 | 4 | 2663 | |||
GenBank mRNA | AB209332.1 | - | 3 | 2404 | ||
AF057034.1 | - | 4 | 887 | |||
AF086735.1 | - | 4 | 1603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
