siRNA Details
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003725.3 | NP_003716.2 | 2 | 260 | ||
XM_005269207.1 | XP_005269264.1 | 3 | 409 | |||
XM_005269208.1 | XP_005269265.1 | 3 | 413 | |||
XM_005269209.1 | XP_005269266.1 | 3 | 400 | |||
XM_006719672.1 | XP_006719735.1 | 3 | 408 | |||
XM_011538925.1 | XP_011537227.1 | 3 | 403 | |||
XM_011538926.1 | XP_011537228.1 | 3 | 393 | |||
XM_011538927.1 | XP_011537229.1 | 3 | 384 | |||
GenBank mRNA | AF016509.1 | - | 1 | 44 | ||
AF223225.1 | - | 2 | 131 | |||
AK313423.1 | - | 2 | 170 | |||
BC020710.1 | - | 2 | 154 | 4700726 | ||
DA086202.1 | - | 3 | 393 | |||
U89281.1 | - | 2 | 149 |
Gene Symbol: RDH16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320108.1 | NP_001307037.1 | 1 | 1956 | ||
NM_003708.4 | NP_003699.3 | 1 | 1956 | |||
GenBank mRNA | AB209332.1 | - | 1 | 1956 | ||
AF057034.1 | - | 1 | 180 | |||
AF086735.1 | - | 1 | 896 | |||
AK311025.1 | - | 3 | 794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
