siRNA Details
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 1 | 189 | ||
NM_023085.3 | NP_075573.2 | 1 | 189 | |||
GenBank mRNA | AB058748.1 | - | 1 | 93 | ||
AF089088.1 | - | 1 | 170 | |||
AF089090.1 | - | 1 | 34 | |||
AF089091.1 | - | 1 | 34 | |||
AF089092.1 | - | 1 | 34 | |||
AF089093.1 | - | 1 | 34 | |||
AF089094.1 | - | 1 | 34 | |||
AF089096.1 | - | 1 | 34 | |||
AK074807.1 | - | 1 | 140 | |||
AK300279.1 | - | 1 | 171 | |||
BC004260.1 | - | 1 | 138 | 3606959 | ||
BC007553.1 | - | 1 | 138 | 2967328 | ||
DA413480.1 | - | 1 | 180 | |||
DA623373.1 | - | 1 | 177 | |||
DB224752.1 | - | 1 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
