siRNA Details
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 1 | 252 | ||
NM_023085.3 | NP_075573.2 | 1 | 252 | |||
GenBank mRNA | AB058748.1 | - | 1 | 156 | ||
AF089088.1 | - | 1 | 233 | |||
AF089090.1 | - | 1 | 97 | |||
AF089091.1 | - | 1 | 97 | |||
AF089092.1 | - | 1 | 97 | |||
AF089093.1 | - | 1 | 97 | |||
AF089094.1 | - | 1 | 97 | |||
AF089096.1 | - | 1 | 97 | |||
AK074807.1 | - | 1 | 203 | |||
AK300279.1 | - | 1 | 234 | |||
AL558905.3 | - | 1 | 250 | |||
BC004260.1 | - | 1 | 201 | 3606959 | ||
BC007553.1 | - | 1 | 201 | 2967328 | ||
DA413480.1 | - | 1 | 243 | |||
DA623373.1 | - | 1 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
